Learn More
Sigma Aldrich Fine Chemicals Biosciences N Methyl D aspartc acd 9250MG
Catalog No. 502902292
Encompass
N-Methyl-D-aspartic acid (NMDA) is an endogenously generated molecule in rat nervous system and endocrine glands. NMDA is present at low level (nmol/g) in the adenohypophysis hypothalamus brain and testis. NMDA is derived from D-Asp by an S-adenosylmethionine-dependent enzyme also referred to as NMDA synthase.
Catalog No. 50-290-2292 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. M3262250MG
N-Methyl-D-aspartic acid (NMDA) is an endogenously generated molecule in rat nervous system and endocrine glands. NMDA is present at low level (nmol/g) in the adenohypophysis hypothalamus brain and testis. NMDA is derived from D-Asp by an S-adenosylmethionine-dependent enzyme also referred to as NMDA synthase.