Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nε-Methyl-L-lysine Hydrochloride 98%, Thermo Scientific™

Catalog No. AC228681000
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC228681000 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC228681000 Supplier Thermo Scientific Chemicals Supplier No. 228681000

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7622-29-9
Molecular Formula C7H17ClN2O2
Molecular Weight (g/mol) 196.68
MDL Number MFCD00012621
InChI Key AQELUQTVJOFFBN-UHFFFAOYNA-N
Synonym h-lys me-oh hcl, h-lys me-oh.hcl, n-methyl-l-lysine hydrochloride, epsilon-n-methyl-l-lysine hcl, n6-methyl-l-lysine hydrochloride, epsilon-n-methyl-l-lysine hydrochloride, 2s-2-amino-6-methylamino hexanoic acid hydrochloride, nepsilon-methyl-l-lysine hydrochloride tlc
PubChem CID 2724391
IUPAC Name hydrogen 2-amino-6-(methylamino)hexanoic acid chloride
SMILES [H+].[Cl-].CNCCCCC(N)C(O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Linear Formula CH3NH(CH2)4CH(NH2)CO2H.HCl
Quantity 100 mg
Beilstein 04, III, 1406
Specific Rotation +18.7° to +20.7° (25°C, 589nm) (c=2, 6N HCl)
Formula Weight 196.68
Percent Purity ≥97.5%
Physical Form Fine Crystalline Powder
Chemical Name or Material Nε-Methyl-L-lysine hydrochloride, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.