Learn More
Cayman Chemical N-methyl Mesoporphyrin IX, Purity: ≥95% (mixture of isomers), CAS: 142234-85-3, 5 mg
SDP
Catalog No. 502940290
Encompass_Preferred
A ferrochelatase inhibitor and turn-on biosensor for DNA; inhibits ferrochelatase in mouse liver mitochondria in vitro; inhibits ferrochelatase in mice at 80 nmol, i.v. leading to porphyrin accumulation in isolated liver mitochondria; has been used as a turn-on biosensor for target DNA sequences when used in complex with a G-quadruplex-forming sequence fused to a DNA sequence complementary to the target sequence; ex/em = 399/610, respectively
Catalog No. 50-294-0290 Supplier Cayman Chemical Supplier No. 272105 MG
A ferrochelatase inhibitor and turn-on biosensor for DNA; inhibits ferrochelatase in mouse liver mitochondria in vitro; inhibits ferrochelatase in mice at 80 nmol, i.v. leading to porphyrin accumulation in isolated liver mitochondria; has been used as a turn-on biosensor for target DNA sequences when used in complex with a G-quadruplex-forming sequence fused to a DNA sequence complementary to the target sequence; ex/em = 399/610, respectively