Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-methyl-n-[3-[4-(phenylmethyl)phenoxy]propyl β-alanine hydrochloride | 423169-68-0 | MFCD00951253 | 99.9% | 363.88 | C20H26ClNO3 | 25 MG
SDP

Catalog No. 5000246199
Encompass_Preferred

SC-57461A is a potent, orally active, nonpeptide inhibitor of leukotriene A4 (LTA4) hydrolase used in research to inhibit leukotriene B4 (LTB4) synthesis and study inflammatory pathways.

  • Potent LTA4 hydrolase inhibition (human recombinant IC50 = 2.5 nM).
  • Cellular activity in whole-blood assays (IC50 ≈ 49 nM).
  • Demonstrated oral efficacy in rodent models.
  • High purity (≈99.9%) and solid, white to off-white appearance.
  • Formula C20H26ClNO3 and molecular weight 363.88 g/mol.
  • Soluble in DMSO (~250 mg/mL); store sealed at 4°C; in solvent store at -80°C for long-term stability.

Catalog No. 50-002-46199 Supplier Medchemexpress LLC Supplier No. HY10322625MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SC-57461A is a potent, orally active, nonpeptide inhibitor of leukotriene A4 (LTA4) hydrolase used in research to inhibit leukotriene B4 (LTB4) synthesis and study inflammatory pathways.

  • Potent LTA4 hydrolase inhibition (human recombinant IC50 = 2.5 nM).
  • Cellular activity in whole-blood assays (IC50 ≈ 49 nM).
  • Demonstrated oral efficacy in rodent models.
  • High purity (≈99.9%) and solid, white to off-white appearance.
  • Formula C20H26ClNO3 and molecular weight 363.88 g/mol.
  • Soluble in DMSO (~250 mg/mL); store sealed at 4°C; in solvent store at -80°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.