Learn More
Medchemexpress LLC N-methyl-N-[3-[4-(phenylmethyl)phenoxy]propyl β-alanine hydrochloride | 423169-68-0 | MFCD00951253 | 99.9% | 363.88 | C20H26ClNO3 | 5 MG
SDP
Catalog No. 5000245967
Encompass_Preferred
SC-57461A is a selective leukotriene A4 hydrolase (LTA4H) inhibitor supplied as the hydrochloride salt for research use. It is provided as a crystalline solid with high stated purity and is used in biochemical and pharmacological assays to probe inflammatory pathways.
  • Selective LTA4 hydrolase inhibition for mechanistic studies.
  • High purity suitable for analytical and pharmacological assays.
  • Available as a solid hydrochloride salt and as a DMSO solution.
  • Molecular weight and formula support accurate dosing and calculations.
  • Stable when stored desiccated at recommended temperatures.
Catalog No. 50-002-45967 Supplier Medchemexpress LLC Supplier No. HY1032265MG
SC-57461A is a selective leukotriene A4 hydrolase (LTA4H) inhibitor supplied as the hydrochloride salt for research use. It is provided as a crystalline solid with high stated purity and is used in biochemical and pharmacological assays to probe inflammatory pathways.
  • Selective LTA4 hydrolase inhibition for mechanistic studies.
  • High purity suitable for analytical and pharmacological assays.
  • Available as a solid hydrochloride salt and as a DMSO solution.
  • Molecular weight and formula support accurate dosing and calculations.
  • Stable when stored desiccated at recommended temperatures.