Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methyl-N-(3-pyridin-4-ylbenzyl)amine, 97%, Thermo Scientific™

Catalog No. CC35646DE
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC35646DE 5 g Amber Glass Bottle
CC35646CB 250 mg Amber Glass Bottle
CC35646DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC35646DE Supplier Thermo Scientific Chemicals Supplier No. CC35646DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 852180-67-7
Molecular Formula C13H14N2
Molecular Weight (g/mol) 198.269
InChI Key PXCMPGTVPUZXRO-UHFFFAOYSA-N
Synonym n-methyl 3-pyridin-4-yl phenyl methanamine, n-methyl-1-3-pyridin-4-yl phenyl methanamine, n-methyl-n-3-pyridin-4-ylbenzyl amine, methyl 3-pyridin-4-yl phenyl methyl amine, n-methyl-1-3-pyridin-4-ylphenyl methanamine, n-methyl-3-pyridin-4-yl benzylamine, n-methyl-n-3-pyridin-4-yl benzylamine, methyl 3-4-pyridyl phenyl methyl amine, 4-3-methylamino methyl phenyl pyridine
PubChem CID 7060574
IUPAC Name N-methyl-1-(3-pyridin-4-ylphenyl)methanamine
SMILES CNCC1=CC=CC(=C1)C2=CC=NC=C2

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 198.27
Percent Purity 95%
Chemical Name or Material N-Methyl-N-(3-pyridin-4-ylbenzyl)amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.