Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methyl-N-(4-pyridylmethyl)amine, 97+%, Thermo Scientific™

Catalog No. MO00914CB
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MO00914CB 250 mg Amber Glass Bottle
MO00914DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MO00914CB Supplier Thermo Scientific Chemicals Supplier No. MO00914CB
Add to Cart

Chemical Identifiers

CAS 6971-44-4
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.171
InChI Key DNBWGFKLIBQQSL-UHFFFAOYSA-N
Synonym n-methyl-1-pyridin-4-yl methanamine, n-methyl-n-4-pyridylmethyl amine, 4-pyridinemethanamine, n-methyl, methylpyridin-4-ylmethylamine, n-methyl-4-pyridylmethylamine, n-methyl-n-4-pyridinylmethyl amine, methyl pyridin-4-ylmethyl amine, n-methylpyridine-4-methylamine, n-methyl-n-pyridin-4-ylmethyl amine, methyl 4-pyridylmethyl amine
PubChem CID 81436
IUPAC Name N-methyl-1-pyridin-4-ylmethanamine
SMILES CNCC1=CC=NC=C1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 122.17
Percent Purity 97%
Chemical Name or Material N-Methyl-N-(4-pyridinylmethyl)amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.