Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methyl-N-[(6-morpholin-4-ylpyridin-2-yl)methyl]amine 90+%, Thermo Scientific™

Catalog No. cc38346cb
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC38346CB 250 mg Amber Glass Bottle
CC38346DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC38346CB Supplier Thermo Scientific Chemicals Supplier No. CC38346CB
Only null left
Add to Cart

Chemical Identifiers

CAS 886851-30-5
Molecular Formula C11H17N3O
Molecular Weight (g/mol) 207.28
MDL Number MFCD08435863
InChI Key CULRQHZBTPUFEB-UHFFFAOYSA-N
Synonym n-methyl-n-6-morpholin-4-ylpyridin-2-yl methyl amine, methyl 6-morpholin-4-yl pyridin-2-yl methyl amine, methyl 6-morpholin-4-yl 2-pyridyl methyl amine, 2-pyridinemethanamine,n-methyl-6-4-morpholinyl, n-methyl-1-6-morpholin-4-yl pyridin-2-yl methanamine
PubChem CID 18525740
SMILES CNCC1=NC(=CC=C1)N1CCOCC1

Specifications

Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name N-methyl-1-(6-morpholin-4-ylpyridin-2-yl)methanamine
Formula Weight 207.28
Percent Purity 90%
Chemical Name or Material N-Methyl-N-[(6-morpholin-4-ylpyridin-2-yl)methyl]amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.