Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methyl-N-nitrosourethane 95.0+%, TCI America™
SDP

Catalog No. N02655G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N02655G 5 g
N026525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. N02655G Supplier TCI America Supplier No. N02655G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 615-53-2
Molecular Formula C4H8N2O3
Molecular Weight (g/mol) 132.12
MDL Number MFCD00013888
InChI Key CAUBWLYZCDDYEF-UHFFFAOYSA-N
Synonym N-Ethoxycarbonyl-N-methylnitrosamine, N-Nitroso-N-methylurethane, Ethyl N-Methyl-N-nitrosocarbamate
PubChem CID 12001
ChEBI CHEBI:82373
IUPAC Name N-methyl-N'-oxoethoxycarbohydrazide
SMILES CCOC(=O)N(C)N=O

Specifications

Boiling Point 66°C
Quantity 5 g
UN Number 2810
Formula Weight 132.12
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Methyl-N-nitrosourethane
RTECSNumber FC6300000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.