Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methyl-n-octylamine 98.0+%, TCI America™
SDP

Catalog No. M22195G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M22195G Supplier TCI America Supplier No. M22195G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2439-54-5
Molecular Formula C9H21N
Molecular Weight (g/mol) 143.274
MDL Number MFCD00048927
InChI Key SEGJNMCIMOLEDM-UHFFFAOYSA-N
Synonym n-methyloctylamine, methyl octyl amine, n-methyl-n-octylamine, 1-octanamine, n-methyl, octylmethylamine, methyloctylamine, unii-4o0o17jz7d, n-methyl octylamine, n-methyl-octylamine, methyl n-octyl amine
PubChem CID 75538
IUPAC Name N-methyloctan-1-amine
SMILES CCCCCCCCNC

Specifications

Color Yellow
Boiling Point 51°C
Quantity 5 g
UN Number 2735
Formula Weight 143.27
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-Methyl-n-octylamine
EINECSNumber (2)-0176
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.