Learn More
N-Methyl-N-(trimethylsilyl)trifluoroacetamide synthesis grade, ≥98%, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| CAS | 24589-78-4 |
| Density | 1.075 g/mL (at 25°C) |
| Boiling Point | 130°C to 132°C |
| Flash Point | 26°C - Closed Cup |
| Packaging | Ampule |
| Linear Formula | CF3 CON(CH3)Si(CH3)3 |
| Quantity | 5 mL |
| Molecular Weight (g/mol) | 199.25 |
| Vapor Density | >1 (vs. air) |
| Formula Weight | 199.25 |
| Show More |
Safety and Handling
| RTECSNumber | AC9800000 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.