Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methyl-N-trimethylsilylacetamide, 98%

Catalog No. AAL0410104
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0410104 2 g
AAL0410109 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0410104 Supplier Thermo Scientific Chemicals Supplier No. L0410104
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 7449-74-3 | C6H15NOSi | 145.277 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7449-74-3
Molecular Formula C6H15NOSi
Molecular Weight (g/mol) 145.277
MDL Number MFCD00040758
InChI Key QHUOBLDKFGCVCG-UHFFFAOYSA-N
Synonym n-methyl-n-trimethylsilyl acetamide, acetamide, n-methyl-n-trimethylsilyl, n-trimethylsilyl-n-methylacetamide, n-methyl-n-tms-acetamide, acmc-1bei6, qhuobldkfgcvcg-uhfffaoysa, n-methylacetamide, tms derivative, n-methyl-n-trimethylsilylacetamide, for gc derivatization gc
PubChem CID 81953
IUPAC Name N-methyl-N-trimethylsilylacetamide
SMILES CC(=O)N(C)[Si](C)(C)C

Specifications

Boiling Point 48°C to 49°C (11mmHg)
Quantity 2 g
UN Number UN1993
Beilstein 506975
Flash Point 48°C (118°F)
Formula Weight 145.28
Percent Purity 95%
Refractive Index 1.439
Sensitivity Moisture sensitive
Density 0.906
Chemical Name or Material N-Methyl-N-trimethylsilylacetamide
Show More Show Less
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 231-217-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.