Learn More
Medchemexpress LLC N-methyl paroxetine | 110429-36-2 | MFCD03788781 | 250mg
SDP
Catalog No. 5000266920
Encompass_Preferred
N-Methylparoxetine is a derivative of Paroxetine that induces Apoptosis NSCLC cells by activating mitogen-activated protein kinase ( MAPK) N-Methylparoxetine has antitumor activity[1]
Catalog No. 50-002-66920 Supplier Medchemexpress LLC Supplier No. HYZ3832250MG
N-Methylparoxetine is a derivative of Paroxetine that induces Apoptosis NSCLC cells by activating mitogen-activated protein kinase ( MAPK) N-Methylparoxetine has antitumor activity[1]