Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-Methyl-phenethylamine

Catalog No. M684235ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M684235ML 5 mL
M6842325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M684235ML Supplier Sigma Aldrich Supplier No. M684235ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • N-methyl-phenethylamine can be used as a reactant:
  • To synthesize n-methyl-phenethylamine based tertiary amines by reacting with different alkyl halides in the presence of triphenylphosphine (tpp) and diisopropylazocarboxylate (diad) via n-alkylation reaction
  • To fabricate photochemically stable, super-sensitive, and highly selective fluorescent film for the detection of n-methamphetamine (an illicit drug)
  • To prepare biologically active squaric acid n-hydroxylamide amide derivatives

Specifications

CAS 589-08-2
Density 0.93 g/mL (at 25°C (literature))
Boiling Point 203°C (lit.)
Molecular Formula C9H13N
Refractive Index n20/D 1.516 (literature)
Linear Formula C6H5CH2CH2NHCH3
MDL Number MFCD00008291
Quantity 5 mL
Synonym N-Methyl-2-phenylethylamine
Molecular Weight (g/mol) 135.21
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.