Learn More
Cayman Chemical N-methyl Protoporphyrin IX, Purity: ≥95% (a mixture of isomers), CAS: 79236-56-9, 1 mg
SDP
Catalog No. 502982998
Encompass_Preferred
A transition state analog that potently inhibits protoporphyrin IX ferrochelatase (Ki = 10 nM), blocking the terminal step of the heme biosynthetic pathway
Catalog No. 50-298-2998 Supplier Cayman Chemical Supplier No. 208461 MG
A transition state analog that potently inhibits protoporphyrin IX ferrochelatase (Ki = 10 nM), blocking the terminal step of the heme biosynthetic pathway