Learn More
Cayman Chemical N-methyl Protoporphyrin IX, Purity: ≥95% (a mixture of isomers), CAS: 79236-56-9, 5 mg
SDP
Catalog No. 502980645
Encompass_Preferred
A transition state analog that potently inhibits protoporphyrin IX ferrochelatase (Ki = 10 nM), blocking the terminal step of the heme biosynthetic pathway
Catalog No. 50-298-0645 Supplier Cayman Chemical Supplier No. 208465 MG
A transition state analog that potently inhibits protoporphyrin IX ferrochelatase (Ki = 10 nM), blocking the terminal step of the heme biosynthetic pathway