Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methyl Pyrrolidone, TARR

Catalog No. 502985656
Encompass
Catalog No. 50-298-5656 Supplier Thermo Scientific Production Chemicals and Services Supplier No. T11038
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

Molecular Weight (g/mol) 99.13 g/mol

Specifications

Boiling Point 395.6 °F
Quantity 4 x 1 gal.
Percent Purity ≥99.5%
Grade Semiconductor
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.