Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methyl-(tetrahydrofuran-3-ylmethyl)amine, 97%, Thermo Scientific™

Catalog No. CC76046CB
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC76046CB 250 mg Amber Glass Bottle
CC76046DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC76046CB Supplier Thermo Scientific Chemicals Supplier No. CC76046CB
Add to Cart

Chemical Identifiers

CAS 7179-93-3
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.176
MDL Number MFCD09040652
InChI Key ZUGRRINNTXSWBR-UHFFFAOYSA-N
Synonym methyl oxolan-3-ylmethyl amine, n-methyl-tetrahydrofuran-3-ylmethyl amine, n-methyl tetrahydrofuran-3-yl methanamine, methyl oxolan-3-yl methyl amine, n-methyl-n-tetrahydrofuran-3-ylmethyl amine, 3-methylamino methyl tetrahydrofuran, methyl-tetrahydrofuran-3-ylmethyl amine, methyl-tetrahydro-furan-3-ylmethyl-amine, n-methyl-1-tetrahydro-3-furanyl methanamine
PubChem CID 16228714
IUPAC Name N-methyl-1-(oxolan-3-yl)methanamine
SMILES CNCC1CCOC1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 115.17
Percent Purity 97%
Chemical Name or Material N-methyl-(tetrahydrofuran-3-ylmethyl)amine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.