Learn More
Medchemexpress LLC N-methylacetamide-d6 | 3669-73-6 | MFCD01073934 | 98.6% | 79.13 | C3HD6NO | 50 MG
SDP
Catalog No. 5000301813
Encompass_Preferred
N-Methylacetamide-d6 is the deuterium-labeled analogue of N-methylacetamide used as a stable isotope standard in analytical and research workflows such as NMR and mass spectrometry. It is provided with supporting analytical documentation and is intended for method development, quantitation, and isotope dilution applications.
  • Deuterium-labeled compound for isotope dilution and internal standard use.
  • High purity suitable for analytical applications.
  • Supplied with certificate of analysis and safety data sheet.
  • Available in multiple small sizes for method development.
  • Stable under recommended storage conditions.
Catalog No. 50-003-01813 Supplier Medchemexpress LLC Supplier No. HYW020018S50MG
Add to Cart
Add to Cart
N-Methylacetamide-d6 is the deuterium-labeled analogue of N-methylacetamide used as a stable isotope standard in analytical and research workflows such as NMR and mass spectrometry. It is provided with supporting analytical documentation and is intended for method development, quantitation, and isotope dilution applications.
  • Deuterium-labeled compound for isotope dilution and internal standard use.
  • High purity suitable for analytical applications.
  • Supplied with certificate of analysis and safety data sheet.
  • Available in multiple small sizes for method development.
  • Stable under recommended storage conditions.