Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methylacetoacetamide (ca. 70% in Water), TCI America™
SDP

Catalog No. M065725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M065725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M065725G Supplier TCI America Supplier No. M065725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 20306-75-6
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.132
MDL Number MFCD00044026
InChI Key ATWLCPHWYPSRBQ-UHFFFAOYSA-N
Synonym n-methylacetoacetamide, n-methyl-3-oxobutyramide, butanamide, n-methyl-3-oxo, n-monomethylacetoacetamide, acetoacet-monomethylamide, unii-pwe9p6v839, n-methyl acetoacetylamide, n-acetoacetylmethylamine, acmc-1chva, butanamide,n-methyl-3-oxo
PubChem CID 88483
IUPAC Name N-methyl-3-oxobutanamide
SMILES CC(=O)CC(=O)NC

Specifications

Color Yellow
Formula Weight 115.13
Quantity 25 g
Physical Form Clear Liquid at 20°C
Chemical Name or Material N-Methylacetoacetamide (ca. 70% in Water)
EINECSNumber (2)-3103
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.