Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-methylalanine | 3913-67-5 | MFCD00037241 | 98.0% | 103.12 g/mol | C4H9NO2 | 50 G
SDP

Catalog No. 5000303125
Encompass_Preferred

H-N-Me-Ala-OH (N-methylalanine) is an N-methylated alanine derivative supplied for research use. It is intended for biochemical and pharmacological studies, including peptide synthesis and investigations of amino-acid-related biological activity. Not for human or clinical use.

  • High purity solid, white to off-white.
  • Molecular weight 103.12 g/mol.
  • Chemical formula C4H9NO2.
  • Supplied in a 50 g amount.
  • Store powder at -20°C for long-term stability.
  • Suitable for peptide synthesis and biochemical research.

Catalog No. 50-003-03125 Supplier Medchemexpress LLC Supplier No. HYW01592650G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

H-N-Me-Ala-OH (N-methylalanine) is an N-methylated alanine derivative supplied for research use. It is intended for biochemical and pharmacological studies, including peptide synthesis and investigations of amino-acid-related biological activity. Not for human or clinical use.

  • High purity solid, white to off-white.
  • Molecular weight 103.12 g/mol.
  • Chemical formula C4H9NO2.
  • Supplied in a 50 g amount.
  • Store powder at -20°C for long-term stability.
  • Suitable for peptide synthesis and biochemical research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.