Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC β-N-methylamino-L-alanine hydrochloride | 16012-55-8 | MFCD00055227 | >=97.0% | 154.60 g/mol | C4H11ClN2O2 | 5 MG
SDP

Catalog No. 5000302935
Encompass_Preferred

β-N-methylamino-L-alanine hydrochloride is a non-proteinogenic, neurotoxic amino acid used as a research reagent in neuroscience and toxicology for studies of excitotoxicity and neurodegeneration. The compound is supplied as the hydrochloride salt, a solid with defined molecular properties for analytical and biological applications.

  • Hydrochloride salt, stable solid form.
  • Cas number 16012-55-8 for unambiguous identification.
  • Molecular weight 154.60 g/mol; formula C4H11ClN2O2.
  • Typical purity ≥97% (depositor-supplied), suitable for analytical and biological research.
  • Applicable for in vitro and in vivo neurotoxicity and mechanistic studies.

Catalog No. 50-003-02935 Supplier Medchemexpress LLC Supplier No. HYW0155465MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

β-N-methylamino-L-alanine hydrochloride is a non-proteinogenic, neurotoxic amino acid used as a research reagent in neuroscience and toxicology for studies of excitotoxicity and neurodegeneration. The compound is supplied as the hydrochloride salt, a solid with defined molecular properties for analytical and biological applications.

  • Hydrochloride salt, stable solid form.
  • Cas number 16012-55-8 for unambiguous identification.
  • Molecular weight 154.60 g/mol; formula C4H11ClN2O2.
  • Typical purity ≥97% (depositor-supplied), suitable for analytical and biological research.
  • Applicable for in vitro and in vivo neurotoxicity and mechanistic studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.