Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-Methylated Pomalidomide

Catalog No. 90149425MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-149-425MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-149-425MG Supplier Sigma Aldrich Supplier No. 90149425MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

N-Methylated pomalidomide is a ligand used as a negative control for pomalidomide (P0018) in the recruitment of the Cereblon (CRBN) protein for targeted protein degradation and PROTAC (proteolysis-targeting chimeras) technology.Technology Spotlight: Degrader Building Blocks for Targeted Protein DegradationPortal: Building PROTAC™ Degraders for Targeted Protein DegradationHijacking the E3 Ubiquitin Ligase Cereblon to Efficiently Target BRD4Targeted Protein Degradation by Small MoleculesSmall-Molecule PROTACS: New Approaches to Protein DegradationPROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

Specifications

CAS 2132978-44-8
Molecular Formula C20H23N3O8
Linear Formula C20H23N3O8
Quantity 50 mg
Synonym 3-(2-(2-((2-(2,6-Dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)amino)ethoxy)ethoxy)propanoic acid; CrosslinkerE3 Ligase ligand conjugate; Protein degrader building block for PROTAC(R) research; Template for synthesis of targeted protein degrader
Molecular Weight (g/mol) 433.41 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.