Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-methylated pomalidomide | 1352827-50-9 | MFCD31703382 | 97.9% | 287.27 g/mol | C14H13N3O4 | 250 MG
SDP

Catalog No. 5000284758
Encompass_Preferred

N-methylated pomalidomide is a research-grade derivative of pomalidomide used in chemical biology and medicinal chemistry. It is a cereblon (CRBN) ligand analog reported to be unable to recruit CRBN, making it suitable as a negative control and as a building block for PROTAC construction.

  • Used as a negative control in cereblon (CRBN) recruitment assays.
  • Suitable as a building block for PROTAC synthesis and linker conjugation.
  • Purity 97.9% (per COA).
  • Molecular formula C14H13N3O4; molecular weight 287.27 g/mol.
  • Store at 4°C and protect from light; in solvent: -80°C for up to 6 months.
  • Available in milligram and gram quantities for synthesis and assay development.

Catalog No. 50-002-84758 Supplier Medchemexpress LLC Supplier No. HYW586822250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-methylated pomalidomide is a research-grade derivative of pomalidomide used in chemical biology and medicinal chemistry. It is a cereblon (CRBN) ligand analog reported to be unable to recruit CRBN, making it suitable as a negative control and as a building block for PROTAC construction.

  • Used as a negative control in cereblon (CRBN) recruitment assays.
  • Suitable as a building block for PROTAC synthesis and linker conjugation.
  • Purity 97.9% (per COA).
  • Molecular formula C14H13N3O4; molecular weight 287.27 g/mol.
  • Store at 4°C and protect from light; in solvent: -80°C for up to 6 months.
  • Available in milligram and gram quantities for synthesis and assay development.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.