Learn More
Medchemexpress LLC N-Methylbenzylamine | 103-67-3 | 121.18 | 1 ML
SDP
Catalog No. 5000460365
Encompass_Preferred
N-Methylbenzylamine is a phenylmethylamine compound that can be found in carrots, suggesting its potential as a biomarker for carrot consumption. It is available as a 10 mM * 1 mL solution in DMSO, ready for reconstitution. This compound is susceptible to spontaneous nitrosation, particularly under acidic conditions.
  • Member of phenylmethylamines
  • Potential biomarker for carrot consumption
  • Available in 10 mM * 1 mL solution in DMSO
  • Susceptible to spontaneous nitrosation
Catalog No. 50-004-60365 Supplier Medchemexpress LLC Supplier No. HYW00742610MM/1ML
N-Methylbenzylamine is a phenylmethylamine compound that can be found in carrots, suggesting its potential as a biomarker for carrot consumption. It is available as a 10 mM * 1 mL solution in DMSO, ready for reconstitution. This compound is susceptible to spontaneous nitrosation, particularly under acidic conditions.
  • Member of phenylmethylamines
  • Potential biomarker for carrot consumption
  • Available in 10 mM * 1 mL solution in DMSO
  • Susceptible to spontaneous nitrosation