Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-Methylbenzylamine | 103-67-3 | 121.18 | 10 MG
SDP

Catalog No. 5000460367
Encompass_Preferred

N-Methylbenzylamine is a phenylmethylamine that can be found in carrots, making it a potential biomarker for the consumption of these food products. It is known to be susceptible to spontaneous nitrosation, a process that is enhanced under acidic conditions.

  • Found in carrots
  • Potential biomarker for carrot consumption
  • Susceptible to spontaneous nitrosation, enhanced under acidic conditions
  • Molecular weight: 121.18
  • CAS number: 103-67-3
  • Purity: 99.55%
  • Store at 4°C, protected from light
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, protected from light

Catalog No. 50-004-60367 Supplier Medchemexpress LLC Supplier No. HYW007426S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Methylbenzylamine is a phenylmethylamine that can be found in carrots, making it a potential biomarker for the consumption of these food products. It is known to be susceptible to spontaneous nitrosation, a process that is enhanced under acidic conditions.

  • Found in carrots
  • Potential biomarker for carrot consumption
  • Susceptible to spontaneous nitrosation, enhanced under acidic conditions
  • Molecular weight: 121.18
  • CAS number: 103-67-3
  • Purity: 99.55%
  • Store at 4°C, protected from light
  • Store in solvent at -80°C for 6 months or -20°C for 1 month, protected from light

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.