Learn More
Medchemexpress LLC N-Methylbenzylamine | 103-67-3 | 121.18 | 100 MG
SDP
Catalog No. 5000460405
Encompass_Preferred
N-Methylbenzylamine is a phenylmethylamine derivative. This compound is naturally found in carrots and is considered a potential biomarker for their consumption. It is also susceptible to spontaneous nitrosation, a process that is enhanced under acidic conditions.
  • Member of phenylmethylamines
  • Found naturally in carrots
  • Potential biomarker for carrot consumption
  • Susceptible to spontaneous nitrosation, enhanced under acidic conditions
Catalog No. 50-004-60405 Supplier Medchemexpress LLC Supplier No. HYW007426100MG
N-Methylbenzylamine is a phenylmethylamine derivative. This compound is naturally found in carrots and is considered a potential biomarker for their consumption. It is also susceptible to spontaneous nitrosation, a process that is enhanced under acidic conditions.
  • Member of phenylmethylamines
  • Found naturally in carrots
  • Potential biomarker for carrot consumption
  • Susceptible to spontaneous nitrosation, enhanced under acidic conditions