Learn More
Medchemexpress LLC N-Methylbenzylamine-d3 | 122025-09-6 | 124.20 | 100 MG
SDP
Catalog No. 5000460366
Encompass_Preferred
N-Methylbenzylamine-d3 is a deuterium-labeled variant of N-Methylbenzylamine, which belongs to the class of phenylmethylamines. This compound has been identified in carrots, suggesting its potential use as a biomarker for the consumption of such food products. Its deuterium labeling makes it suitable for specific research applications, including tracing and quantitative analysis using techniques like NMR, GC-MS, or LC-MS.The features and benefits of N-Methylbenzylamine-d3 include:
  • It is a deuterium labeled compound.
  • It can be found in carrots.
  • It serves as a potential biomarker for carrot consumption.
  • It is suitable for research use only.
  • It has applications as a tracer.
  • It can be used as an internal standard for quantitative analysis.
  • Its deuterium substitution can impact pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-60366 Supplier Medchemexpress LLC Supplier No. HYW007426S100MG
N-Methylbenzylamine-d3 is a deuterium-labeled variant of N-Methylbenzylamine, which belongs to the class of phenylmethylamines. This compound has been identified in carrots, suggesting its potential use as a biomarker for the consumption of such food products. Its deuterium labeling makes it suitable for specific research applications, including tracing and quantitative analysis using techniques like NMR, GC-MS, or LC-MS.The features and benefits of N-Methylbenzylamine-d3 include:
  • It is a deuterium labeled compound.
  • It can be found in carrots.
  • It serves as a potential biomarker for carrot consumption.
  • It is suitable for research use only.
  • It has applications as a tracer.
  • It can be used as an internal standard for quantitative analysis.
  • Its deuterium substitution can impact pharmacokinetic and metabolic profiles of drugs.