Learn More
Medchemexpress LLC N-methylbenzylamine-d3 | 122025-09-6 | 124.20 | 5 MG
SDP
Catalog No. 5000460368
Encompass_Preferred
N-Methylbenzylamine-d3 is a deuterium-labeled N-Methylbenzylamine, belonging to the phenylmethylamines. It can be found in carrots, making it a potential biomarker for the consumption of such food products. This compound is intended for research use only, not for patient use.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration may affect pharmacokinetic and metabolic profiles.
Catalog No. 50-004-60368 Supplier Medchemexpress LLC Supplier No. HYW007426S5MG
N-Methylbenzylamine-d3 is a deuterium-labeled N-Methylbenzylamine, belonging to the phenylmethylamines. It can be found in carrots, making it a potential biomarker for the consumption of such food products. This compound is intended for research use only, not for patient use.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration may affect pharmacokinetic and metabolic profiles.