Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methylcyclohexylamine, 98%

Catalog No. AAL13872AE
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL13872AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL13872AE Supplier Thermo Scientific Chemicals Supplier No. L13872AE

missing translation for 'validMainframeMsg'

CAS: 100-60-7 | C7H15N | 113.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-60-7
Molecular Formula C7H15N
Molecular Weight (g/mol) 113.20
MDL Number MFCD00003832
InChI Key XTUVJUMINZSXGF-UHFFFAOYSA-N
Synonym n-methylcyclohexylamine, methylcyclohexylamine, n-cyclohexylmethylamine, cyclohexanamine, n-methyl, n-methyl-n-cyclohexylamine, methylamino cyclohexane, cyclohexylamine, n-methyl, cyclohexyl methyl amine, n-cyclohexyl-n-methylamine, unii-yh8pf4c1ow
PubChem CID 7514
ChEBI CHEBI:59018
IUPAC Name N-methylcyclohexanamine
SMILES CNC1CCCCC1

Specifications

Melting Point -8°C
Density 0.857
Boiling Point 149°C
Flash Point 35°C (95°F)
Refractive Index 1.456
Quantity 100 mL
UN Number UN2734
Beilstein 1523664
Sensitivity Air Sensitive
Formula Weight 113.2
Percent Purity 98%
Chemical Name or Material N-Methylcyclohexylamine
Show More Show Less
Hazard Category H226-H290-H302-H312-H314
Hazard Statement GHS H Statement
H301-H314-H318-H226-H332
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if inhaled.
Precautionary Statement P210-P233-P234-P240-P243-P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P374-P380-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, FLAMMABLE, N.O.S.
EINECSNumber 202-869-3
RTECSNumber GX1529000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.