Learn More
Cayman Chemical N-Methylcytisine, Purity: ≥95%, CAS: 486-86-2, 100 mg
SDP
Catalog No. 502910244
Encompass_Preferred
An alkaloid with diverse biological activities; an nAChR agonist; selectively binds to nAChRs over mAChRs (IC50s = 0.05 and 417 µM, respectively); elicits whole-cell currents in Xenopus oocytes expressing α4β2, α4β4, and α7 subunit-containing nAChRs (EC50s = 13, 13, and 340 µM, respectively); nematocidal against B. xylophilus; inhibits LPS-induced NO production in RAW 264.7 macrophages (IC50 = 33.3 µM) and proliferation of SMMC-7721 cells at 4 mg/ml
Catalog No. 50-291-0244 Supplier Cayman Chemical Supplier No. 34752100 MG
An alkaloid with diverse biological activities; an nAChR agonist; selectively binds to nAChRs over mAChRs (IC50s = 0.05 and 417 µM, respectively); elicits whole-cell currents in Xenopus oocytes expressing α4β2, α4β4, and α7 subunit-containing nAChRs (EC50s = 13, 13, and 340 µM, respectively); nematocidal against B. xylophilus; inhibits LPS-induced NO production in RAW 264.7 macrophages (IC50 = 33.3 µM) and proliferation of SMMC-7721 cells at 4 mg/ml