Learn More
Medchemexpress LLC HY-N0443 5mg Medchemexpress, N-Methylcytisine CAS:486-86-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN04435MG
Encompass_Preferred
Medchemexpress, HY-N0443 5mg N-Methylcytisine CAS:486-86-2 N-Methylcytisine (Caulophylline), a tricyclic quinolizidine alkaloid, exerts hypoglycaemic, analgesic and anti-inflammatory activities. N-methylcytisine is a selective ligand of nicotinic receptors of acetylcholine in the central nervous system and has a high affinity (Kd = 50 nM) to nicotinic acetylcholine receptors (nAChR) from squid optical ganglia. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2790