Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-Methyldiethanolamine

Catalog No. 471828250ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
250 mL
2 L
18 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-182-8250ML 250 mL
47-182-818L 18 L
47-182-825ML 25 mL
47-182-82L 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 47-182-8250ML Supplier Sigma Aldrich Supplier No. 471828250ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • N-methyldiethanolamine is an alkanolamine.n-methyldiethanolamine (mdea) may be employed in the following studies:
  • Synthesis of aluminophosphate-based molecular sieves
  • Preparation of n-methyl-n-r-n,n-bis(2-hydroxyethyl) ammonium bromides
  • Preparation of cationic polyurethane dispersions

Specifications

CAS 105-59-9
Density 1.038 g/mL (at 25°C (literature))
Boiling Point 246°C to 248°C (lit.)
Molecular Formula C5H13NO2
Linear Formula CH3N(CH2CH2OH)2
Refractive Index n20/D 1.469 (literature)
MDL Number MFCD00002848
Quantity 250 mL
Synonym 2,2 -Methyliminodiethanol; N,N-Bis(2-hydroxyethyl)methylamine; MDEA
Molecular Weight (g/mol) 119.16
Percent Purity ≥99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.