Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methyldiphenylamine, 97%

Catalog No. AC414210250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC414210250 25 g Glass bottle
AC414210050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC414210250 Supplier Thermo Scientific Chemicals Supplier No. 414210250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 552-82-9 | C13H13N | 183.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 552-82-9
Molecular Formula C13H13N
Molecular Weight (g/mol) 183.25
MDL Number MFCD00041900
InChI Key DYFFAVRFJWYYQO-UHFFFAOYSA-N
Synonym n-methyldiphenylamine, benzenamine, n-methyl-n-phenyl, methyldiphenylamine, n,n-diphenylmethylamine, diphenylamine, n-methyl, unii-b28zgh99ih, diphenyl methylamine, n-methyl-n-phenyl-aniline, b28zgh99ih, methyldiphenylamin
PubChem CID 11098
IUPAC Name N-methyl-N-phenylaniline
SMILES CN(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color White to Beige
Density 1.05g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.6215 to 1.6235 (20°C, 589nm)
Linear Formula C6H5N(CH3)C6H5
Quantity 25 g
Merck Index 15, 6126
Formula Weight 183.25
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-Methyldiphenylamine, 97%
Show More Show Less
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 209-023-2
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.