Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methylformamide, 99%

Catalog No. p-163534
Change view
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC414331000 100 g Glass bottle
AC414330010 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC414331000 Supplier Thermo Scientific Chemicals Supplier No. 414331000
Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 123-39-7 | C2H5NO | 59.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-39-7
Molecular Formula C2H5NO
Molecular Weight (g/mol) 59.06
MDL Number MFCD00003280
InChI Key ATHHXGZTWNVVOU-UHFFFAOYSA-N
Synonym methylformamide, monomethylformamide, formamide, n-methyl, n-formylmethylamine, n-methyl-formamide, n-monomethylformamide, n-methyl formamide, hconhch3, unii-xpe4g7y986, formamide, methyl
PubChem CID 31254
ChEBI CHEBI:7438
IUPAC Name N-methylformamide
SMILES CNC=O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -3.2°C
Color Colorless
Density 1.0030g/mL
Boiling Point 199.0°C to 201.0°C
Flash Point 111°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4310 to 1.4330
Linear Formula HCONHCH3
Quantity 100 g
Beilstein 04, 58
Specific Gravity 1.003
Merck Index 15, 6147
Solubility Information Solubility in water: miscible. Other solubilities: soluble in acetone and alcohol
Viscosity 1.9 mPa.s (20°C)
Formula Weight 59.06
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N-Methylformamide, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful in contact with skin.
May damage the unborn child.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF exposed o
EINECSNumber 204-624-6
RTECSNumber LQ3000000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.