Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich N-Methylfulleropyrrolidine

Catalog No. 668184100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
66-818-4100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 66-818-4100MG Supplier Sigma Aldrich Supplier No. 668184100MG
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Photoinduced energy and electron transfer between oligothienylenevinylenes and N-methylfulleropyrrolidine (MP-C60) has been studied in details. MP-C60 acts as an electron acceptor.Soluble functionalized fullerene. Acts as electron acceptor in mixtures or when linked toconjugated molecules and polymers. Used as spectroscopic and redox reference for C60-based molecular heterojunctions.

Specifications

CAS 151872-44-5
Molecular Formula C63H7N
Linear Formula C63H7N
Quantity 100 mg
Synonym MFP; MPC60; NMPC60
Molecular Weight (g/mol) 777.74
Percent Purity 99% (HPLC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.