Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methylfurohydroxamic Acid 99.0+%, TCI America™
SDP

Catalog No. A50825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A50825G 5 g
A50821G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A50825G Supplier TCI America Supplier No. A50825G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 109531-96-6
Molecular Formula C6H7NO3
Molecular Weight (g/mol) 141.126
MDL Number MFCD00144939
InChI Key KCAHIEOWHDMBKM-UHFFFAOYSA-N
Synonym N-(2-Furoyl)-N-methylhydroxylamine, N-MFHA
PubChem CID 565703
IUPAC Name N-hydroxy-N-methylfuran-2-carboxamide
SMILES CN(C(=O)C1=CC=CO1)O

Specifications

Melting Point 101°C
Color White
Quantity 5 g
Formula Weight 141.13
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-Methylfurohydroxamic Acid [Chelating Reagent for HPLC]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.