Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-methylhistamine dihydrochloride | 16503-22-3 | MFCD00134838 | 198.09 g/mol | C6H13Cl2N3 | 5 MG
SDP

Catalog No. 5000211981
Encompass_Preferred

N-Methylhistamine dihydrochloride is a research-grade histamine H3 receptor agonist used in pharmacological and biochemical studies of histamine signaling and mast cell-related research. It is supplied as a solid salt for laboratory use and intended for research purposes only.

  • Potent agonist of the histamine H3 receptor.
  • White to off-white solid.
  • Molecular formula C6H13Cl2N3; molecular weight 198.09 g/mol.
  • CAS number 16503-22-3 for unambiguous identification.
  • Available in small research pack sizes, including 5 mg.
  • Store sealed at 4°C; in solution store -80°C up to 6 months.

Catalog No. 50-002-11981 Supplier Medchemexpress LLC Supplier No. HY1010055MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-Methylhistamine dihydrochloride is a research-grade histamine H3 receptor agonist used in pharmacological and biochemical studies of histamine signaling and mast cell-related research. It is supplied as a solid salt for laboratory use and intended for research purposes only.

  • Potent agonist of the histamine H3 receptor.
  • White to off-white solid.
  • Molecular formula C6H13Cl2N3; molecular weight 198.09 g/mol.
  • CAS number 16503-22-3 for unambiguous identification.
  • Available in small research pack sizes, including 5 mg.
  • Store sealed at 4°C; in solution store -80°C up to 6 months.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.