Learn More
Selleck Chemical LLC N-Methylhydantoin
SDP
Catalog No. 5000003881
Encompass_Preferred
N-Methylhydantoin (1-Methylhydantoin Dioxy-creatinine) is a small molecular weight polar substance the product of degradation of creatinine by bacteria It is an unexpected metabolite of the intelligence-affecting substance dupracetam
Catalog No. 50-000-03881 Supplier Selleck Chemical LLC Supplier No. S618425MG
N-Methylhydantoin (1-Methylhydantoin Dioxy-creatinine) is a small molecular weight polar substance the product of degradation of creatinine by bacteria It is an unexpected metabolite of the intelligence-affecting substance dupracetam