Learn More
Medchemexpress LLC N-Methylhydantoin | 616-04-6 | 99.9% | 114.10 | 50 G
SDP
Catalog No. 5000436721
Encompass_Preferred
N-Methylhydantoin is a product of degradation of creatinine by bacteria.
  • High purity chemical, typically 99.9% pure.
  • Appears as a solid, white to off-white in color.
  • Soluble in DMSO for in vitro applications.
  • Can be prepared for in vivo studies using various protocols.
Catalog No. 50-004-36721 Supplier Medchemexpress LLC Supplier No. HY11338250G
Add to Cart
Add to Cart
N-Methylhydantoin is a product of degradation of creatinine by bacteria.
  • High purity chemical, typically 99.9% pure.
  • Appears as a solid, white to off-white in color.
  • Soluble in DMSO for in vitro applications.
  • Can be prepared for in vivo studies using various protocols.