Learn More
Medchemexpress LLC N-Methyliminodiacetic acid | 4408-64-4 | 147.13 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0078725G
Encompass_Preferred
N-Methyliminodiacetic acid is a biochemical reagent (2,2'-(Methylazanediyl)diacetic acid) for life science research. It is intended for research use only and may cause skin, eye, and respiratory irritation. Appropriate protective measures should be taken in a well-ventilated area.
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • Causes skin irritation
  • Causes serious eye irritation
  • May cause respiratory irritation
Catalog No. 50-004-46519