Learn More
Medchemexpress LLC N-Methylindan-2-amine hydrochloride | 10408-85-2 | 99.9% | 183.68 | 1 G
SDP
Catalog No. 5000435992
Encompass_Preferred
N-Methylindan-2-amine hydrochloride (compound 3b) is an N-alkylated congener of phenylethylamine that increases hot plate reaction time in mice without inducing dopaminergic effects. It can be used to study central noradrenergic mechanisms.
  • Increases hot plate reaction time in mice without inducing dopaminergic effects.
  • Can be used to study central noradrenergic mechanisms.
  • For research use only.
Catalog No. 50-004-35992 Supplier Medchemexpress LLC Supplier No. HY1138331G
N-Methylindan-2-amine hydrochloride (compound 3b) is an N-alkylated congener of phenylethylamine that increases hot plate reaction time in mice without inducing dopaminergic effects. It can be used to study central noradrenergic mechanisms.
  • Increases hot plate reaction time in mice without inducing dopaminergic effects.
  • Can be used to study central noradrenergic mechanisms.
  • For research use only.