Learn More
Medchemexpress LLC N-Methylindan-2-amine hydrochloride | 10408-85-2 | 99.9% | 183.68 | 5 G
SDP
Catalog No. 5000438020
Encompass_Preferred
N-Methylindan-2-amine hydrochloride (compound 3b) is an N-alkylated congener of phenylethylamine that increases hot plate reaction time in mice without inducing dopaminergic effects. It can be used to study central noradrenergic mechanisms. This product is for research use only and not sold to patients.
  • Used to study central noradrenergic mechanisms
  • Increases hot plate reaction time in mice
  • Does not induce dopaminergic effects
Catalog No. 50-004-38020 Supplier Medchemexpress LLC Supplier No. HY1138335G
N-Methylindan-2-amine hydrochloride (compound 3b) is an N-alkylated congener of phenylethylamine that increases hot plate reaction time in mice without inducing dopaminergic effects. It can be used to study central noradrenergic mechanisms. This product is for research use only and not sold to patients.
  • Used to study central noradrenergic mechanisms
  • Increases hot plate reaction time in mice
  • Does not induce dopaminergic effects