Learn More
Medchemexpress LLC HY-114758 50mg Medchemexpress, Pyridoxal isonicotinoyl hydrazone CAS:737-86-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11475850MG
Encompass_Preferred
Medchemexpress, HY-18407 100mg N-Methylisatoic anhydride CAS:10328-92-4 N-Methylisatoic anhydride (NMIA) is a 2'-OH selective acylation agent of RNAs, and is widely used forresolving secondary RNA structures using the SHAPE (Selective 2'-Hydroxyl Acylation Analyzed by Primer Extension) technology. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9373