Learn More
Medchemexpress LLC N-methylmethanesulfonamide chromanol derivative | 223749-46-0 | MFCD12756323 | 100.0% | 411.44 g·mol⁻¹ | C17H24F3NO5S | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10636950MG
Encompass_Preferred
HMR 1556 is a chromanol-derived research compound that potently and selectively blocks the slowly activating delayed rectifier potassium current (IKs). It is used as a pharmacological tool in cardiac electrophysiology and pharmacology to probe IKs-mediated repolarization in vitro and in vivo. Reported IC50 values include 10.5 nM (canine left ventricular myocytes) and 34 nM (guinea pig left ventricular myocytes).
  • Potent IKs blockade at low-nanomolar concentrations.
  • Selective for IKs with limited off-target activity at pharmacologically relevant doses.
  • Suitable for in vitro and in vivo electrophysiology and pharmacology studies.
  • High-purity solid form, soluble in DMSO for stock solutions.
  • Documented solubility and storage conditions for reliable experimental use.
Catalog No. 50-002-42473