Learn More
Medchemexpress LLC N-Methylmoranoline | 69567-10-8 | 177.20 | 10 MG
SDP
Catalog No. 5000446791
Encompass_Preferred
N-Methylmoranoline is an alpha-glucosidase inhibitor. It appears as a white to off-white solid.
  • Functions as an alpha-glucosidase inhibitor
  • Suitable as a reference compound
  • Powder stable for 3 years at -20°C
  • In solvent stable for 6 months at -80°C, or 1 month at -20°C
Catalog No. 50-004-46791 Supplier Medchemexpress LLC Supplier No. HYU0009010MG
N-Methylmoranoline is an alpha-glucosidase inhibitor. It appears as a white to off-white solid.
  • Functions as an alpha-glucosidase inhibitor
  • Suitable as a reference compound
  • Powder stable for 3 years at -20°C
  • In solvent stable for 6 months at -80°C, or 1 month at -20°C