Learn More
Apexbio Technology LLC N-Methylnicotinamide 114-33-0 200mg
SDP
Catalog No. 5000088419
Encompass_Preferred
N-Methylnicotinamide (CAS 114-33-0) is a methylated derivative of nicotinamide and a primary metabolic product of niacinamide and niacin (vitamin B3) catabolism It does not have a defined therapeutic target or approved clinical use but plays a significant role in biomedical research as an indicator of vitamin B3 status Measurement of urinary N-methylnicotinamide is routinely employed to assess niacin sufficiency and diagnose deficiencies reflecting metabolic function related to NAD/NADP biosynthesis pathways This compound thus serves as a valuable biomarker in nutritional and metabolic research contexts
Catalog No. 50-000-88419 Supplier Apexbio Technology LLC Supplier No. C6682200
N-Methylnicotinamide (CAS 114-33-0) is a methylated derivative of nicotinamide and a primary metabolic product of niacinamide and niacin (vitamin B3) catabolism It does not have a defined therapeutic target or approved clinical use but plays a significant role in biomedical research as an indicator of vitamin B3 status Measurement of urinary N-methylnicotinamide is routinely employed to assess niacin sufficiency and diagnose deficiencies reflecting metabolic function related to NAD/NADP biosynthesis pathways This compound thus serves as a valuable biomarker in nutritional and metabolic research contexts