Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC N-methylnicotinamide | 114-33-0 | MFCD00006384 | 99.6% | 136.15 g/mol | C7H8N2O | 10MM 1ML
SDP

Catalog No. 5000239042
Encompass_Preferred

N-methylnicotinamide is an endogenous metabolite supplied for research and analytical use. It is available as a ready-to-use solution (10 mM in DMSO, 1 mL) and in multiple solid pack sizes to support analytical standards and metabolic studies.

  • Provides a 10 mM solution in DMSO for immediate use
  • High purity: 99.6%
  • CAS number: 114-33-0
  • Chemical formula: C7H8N2O
  • Molecular weight: 136.15 g/mol
  • Suitable for analytical standards and metabolic studies
  • Available in multiple solid pack sizes for bulk needs

Catalog No. 50-002-39042 Supplier Medchemexpress LLC Supplier No. HY12412410MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N-methylnicotinamide is an endogenous metabolite supplied for research and analytical use. It is available as a ready-to-use solution (10 mM in DMSO, 1 mL) and in multiple solid pack sizes to support analytical standards and metabolic studies.

  • Provides a 10 mM solution in DMSO for immediate use
  • High purity: 99.6%
  • CAS number: 114-33-0
  • Chemical formula: C7H8N2O
  • Molecular weight: 136.15 g/mol
  • Suitable for analytical standards and metabolic studies
  • Available in multiple solid pack sizes for bulk needs

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.