Learn More
Selleck Chemical LLC N-Methylnicotinamide S3321-100mg
SDP
Catalog No. 503109101
Encompass_Preferred
N-Methylnicotinamide (MNA Nicotinyl Methylamide N-Methyl-3-pyridinecarboxamide) is an active endogenous metabolite that improves endothelial dysfunction and attenuates atherogenesis via the modulation of ADMA-DDAH axis
Catalog No. 50-310-9101 Supplier Selleck Chemical LLC Supplier No. S3321100MG
N-Methylnicotinamide (MNA Nicotinyl Methylamide N-Methyl-3-pyridinecarboxamide) is an active endogenous metabolite that improves endothelial dysfunction and attenuates atherogenesis via the modulation of ADMA-DDAH axis