Learn More
Cayman Chemical N-Methylnuciferine (iodide), Purity: ≥98%, CAS: 53098-68-3, 10 mg
SDP
Catalog No. 502942311
Encompass_Preferred
An aporphine alkaloid with diverse biological activities; scavenges DPPH radicals (IC50 = 711.76 µg/ml); inhibits α-glucosidase and α-amylase (IC50s = 2,627.78 and 1,818.18 µg/ml, respectively); reduces LPS-induced NO production in BV-2 microglia at 20 µM
Catalog No. 50-294-2311 Supplier Cayman Chemical Supplier No. 4027610 MG
An aporphine alkaloid with diverse biological activities; scavenges DPPH radicals (IC50 = 711.76 µg/ml); inhibits α-glucosidase and α-amylase (IC50s = 2,627.78 and 1,818.18 µg/ml, respectively); reduces LPS-induced NO production in BV-2 microglia at 20 µM